Assay Details
| Gene Symbol: TPM3-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TPM3-PDGFRB_T7P11.0 | - | 45 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPM3-PDGFRB fusion| Gene Symbol: TPM3-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TPM3-PDGFRB_T7P11.0 | - | 45 | 91 | ||