Assay Details
| Gene Symbol: WHSC1L1-NUTM1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_WHSC1L1-NUTM1_W7N2.0 | - | 63 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WHSC1L1-NUTM1 fusion| Gene Symbol: WHSC1L1-NUTM1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_WHSC1L1-NUTM1_W7N2.0 | - | 63 | 75 | ||