Assay Details
| Gene Symbol: ZKSCAN1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ZKSCAN1-MET_Z3M15.0 | - | 64 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZKSCAN1-MET fusion| Gene Symbol: ZKSCAN1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ZKSCAN1-MET_Z3M15.0 | - | 64 | 88 | ||