Assay Details
| Gene Symbol: AKAP13-NTRK3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AKAP13-NTRK3_A14N14.0 | - | 90 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AKAP13-NTRK3 fusion| Gene Symbol: AKAP13-NTRK3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AKAP13-NTRK3_A14N14.0 | - | 90 | 103 | ||
Set Membership: |
|