Assay Details
| Gene Symbol: AR-AR fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AR-AR_A3A3int4_V9.0 | - | 65 | 83 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AR-AR fusion| Gene Symbol: AR-AR fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AR-AR_A3A3int4_V9.0 | - | 65 | 83 | ||