Assay Details
| Gene Symbol: ATP1B1-PRKACA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ATP1B1-PRKACA_A1P2.0 | - | 50 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP1B1-PRKACA fusion| Gene Symbol: ATP1B1-PRKACA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ATP1B1-PRKACA_A1P2.0 | - | 50 | 103 | ||
Set Membership: |
|