Assay Details
| Gene Symbol: BIRC6-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BIRC6-ALK_B10A20.0 | - | 58 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BIRC6-ALK fusion| Gene Symbol: BIRC6-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BIRC6-ALK_B10A20.0 | - | 58 | 76 | ||