Assay Details
| Gene Symbol: BRD3-NUTM1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BRD3-NUTM1_B10N2.0 | - | 43 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRD3-NUTM1 fusion| Gene Symbol: BRD3-NUTM1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BRD3-NUTM1_B10N2.0 | - | 43 | 80 | ||