Assay Details
| Gene Symbol: BRD4-NUTM1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BRD4-NUTM1_B14N2del585.0 | - | 89 | 77 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRD4-NUTM1 fusion| Gene Symbol: BRD4-NUTM1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BRD4-NUTM1_B14N2del585.0 | - | 89 | 77 | ||