Assay Details
| Gene Symbol: CLIP1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CLIP1-ALK_C13A20.0 | - | 43 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLIP1-ALK fusion| Gene Symbol: CLIP1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CLIP1-ALK_C13A20.0 | - | 43 | 93 | ||