Assay Details
| Gene Symbol: CLTC-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CLTC-ALK_C31A20_COSF470.0 | - | 82 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLTC-ALK fusion| Gene Symbol: CLTC-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CLTC-ALK_C31A20_COSF470.0 | - | 82 | 100 | ||