Assay Details
| Gene Symbol: DCTN1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_DCTN1-ALK_D29A20.0 | - | 50 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCTN1-ALK fusion| Gene Symbol: DCTN1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_DCTN1-ALK_D29A20.0 | - | 50 | 100 | ||