Assay Details
| Gene Symbol: DTD1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_DTD1-PDGFRB_D4P12.0 | - | 50 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DTD1-PDGFRB fusion| Gene Symbol: DTD1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_DTD1-PDGFRB_D4P12.0 | - | 50 | 86 | ||