Assay Details
| Gene Symbol: ERBB2-GRB7 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ERBB2-GRB7_E29G10.0 | - | 49 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERBB2-GRB7 fusion| Gene Symbol: ERBB2-GRB7 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ERBB2-GRB7_E29G10.0 | - | 49 | 87 | ||