Assay Details
| Gene Symbol: FGFR2-AHCYL1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-AHCYL1_F17A2.0 | - | 87 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR2-AHCYL1 fusion| Gene Symbol: FGFR2-AHCYL1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-AHCYL1_F17A2.0 | - | 87 | 97 | ||