Assay Details
| Gene Symbol: FGFR2-NOL4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-NOL4_F17N7.0 | - | 83 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR2-NOL4 fusion| Gene Symbol: FGFR2-NOL4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-NOL4_F17N7.0 | - | 83 | 93 | ||