Assay Details
| Gene Symbol: FNDC3B-PIK3CA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FNDC3B-PIK3CA_F3P2.0 | - | 90 | 111 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FNDC3B-PIK3CA fusion| Gene Symbol: FNDC3B-PIK3CA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FNDC3B-PIK3CA_F3P2.0 | - | 90 | 111 | ||
Set Membership: |
|