Assay Details
| Gene Symbol: GNAI1-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GNAI1-BRAF_G1B10_COSF1442.0 | - | 84 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNAI1-BRAF fusion| Gene Symbol: GNAI1-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GNAI1-BRAF_G1B10_COSF1442.0 | - | 84 | 90 | ||