Assay Details
| Gene Symbol: LMNA-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_LMNA-NTRK1_L10N11.0 | - | 49 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LMNA-NTRK1 fusion| Gene Symbol: LMNA-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_LMNA-NTRK1_L10N11.0 | - | 49 | 97 | ||