Assay Details
| Gene Symbol: MYBL1-YTHDF3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYBL1-YTHDF3_M8Y4int3.0 | - | 76 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYBL1-YTHDF3 fusion| Gene Symbol: MYBL1-YTHDF3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYBL1-YTHDF3_M8Y4int3.0 | - | 76 | 95 | ||