Assay Details
| Gene Symbol: MYB-NFIB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYB-NFIB_M15N11_COSF880.0 | - | 85 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYB-NFIB fusion| Gene Symbol: MYB-NFIB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYB-NFIB_M15N11_COSF880.0 | - | 85 | 93 | ||