Assay Details
| Gene Symbol: MYH13-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYH13-RET_M35R12.0 | - | 82 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYH13-RET fusion| Gene Symbol: MYH13-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYH13-RET_M35R12.0 | - | 82 | 95 | ||