Assay Details
| Gene Symbol: MYO18A-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYO18A-PDGFRB_M41P10.0 | - | 51 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYO18A-PDGFRB fusion| Gene Symbol: MYO18A-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYO18A-PDGFRB_M41P10.0 | - | 51 | 97 | ||