Assay Details
| Gene Symbol: NTRK1-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NTRK1-NTRK1_N6N8.0 | - | 85 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NTRK1-NTRK1 fusion| Gene Symbol: NTRK1-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NTRK1-NTRK1_N6N8.0 | - | 85 | 95 | ||
Set Membership: |
|