Assay Details
| Gene Symbol: NUP214-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NUP214-BRAF_N21B10.0 | - | 53 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUP214-BRAF fusion| Gene Symbol: NUP214-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NUP214-BRAF_N21B10.0 | - | 53 | 91 | ||