Assay Details
| Gene Symbol: SLC3A2-NRG1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SLC3A2-NRG1_S5N6.0 | - | 89 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC3A2-NRG1 fusion| Gene Symbol: SLC3A2-NRG1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SLC3A2-NRG1_S5N6.0 | - | 89 | 85 | ||