Assay Details
| Gene Symbol: TFG-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TFG-ALK_T7A19.0 | - | 51 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TFG-ALK fusion| Gene Symbol: TFG-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TFG-ALK_T7A19.0 | - | 51 | 95 | ||