Assay Details
| Gene Symbol: TMPRSS2-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TMPRSS2-BRAF_T3B11.0 | - | 52 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS2-BRAF fusion| Gene Symbol: TMPRSS2-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TMPRSS2-BRAF_T3B11.0 | - | 52 | 91 | ||