Assay Details
| Gene Symbol: TRIM33-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TRIM33-RET_T11R12.0 | - | 56 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM33-RET fusion| Gene Symbol: TRIM33-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TRIM33-RET_T11R12.0 | - | 56 | 96 | ||