Assay Details
| Gene Symbol: CAPZA2-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CAPZA2-MET_C1M6.0 | - | 60 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAPZA2-MET fusion| Gene Symbol: CAPZA2-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CAPZA2-MET_C1M6.0 | - | 60 | 99 | ||