Assay Details
| Gene Symbol: MSN-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MSN-ALK_M11int12A20.0 | - | 103 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MSN-ALK fusion| Gene Symbol: MSN-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MSN-ALK_M11int12A20.0 | - | 103 | 102 | ||
Set Membership: |
|