Assay Details
| Gene Symbol: PRKCZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011541775.2 | XP_011540077.1 | 11 | 1188 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKCZ| Gene Symbol: PRKCZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011541775.2 | XP_011540077.1 | 11 | 1188 | 76 | |
Set Membership: |
|