Assay Details
| Gene Symbol: NPPA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037806.1 | 4 | 1541 | 85 | ||
| GenBank mRNA | BQ776822.1 | - | 3 | 396 | 85 | |
| CA773133.1 | - | 3 | 411 | 85 | ||
| FJ706070.1 | - | 4 | 1541 | 85 | ||
| FJ706071.1 | - | 4 | 1418 | 85 | ||
| FJ706078.1 | - | 2 | 238 | 85 | ||
More Information
Set Membership: |
|

NPPA-AS1
Genomic Map