Assay Details
| Gene Symbol: HS6ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004807.2 | NP_004798.3 | 2 | 3824 | 119 | |
| GenBank mRNA | AK058121.1 | - | 1 | 1648 | 119 | |
| AL831893.1 | - | 1 | 1552 | 119 | ||
| BC001196.1 | - | 1 | 3073 | 119 | ||
| BC047087.1 | - | 1 | 967 | 119 | ||
More Information
Set Membership: |
|

HS6ST1
Genomic Map