Assay Details
| Gene Symbol: UPF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023011.3 | NP_075387.1 | 10 | 2165 | 136 | |
| NM_080687.2 | NP_542418.1 | 9 | 2066 | 136 | ||
| XM_005266202.4 | XP_005266259.1 | 7 | 1791 | 136 | ||
| XM_006719991.3 | XP_006720054.1 | 9 | 2040 | 136 | ||
| XM_011534844.1 | XP_011533146.1 | 8 | 1933 | 136 | ||
| XM_011534845.2 | XP_011533147.1 | 10 | 2693 | 136 | ||
| XM_011534847.2 | XP_011533149.1 | 9 | 2505 | 136 | ||
| XM_011534848.2 | XP_011533150.1 | 10 | 2650 | 136 | ||
| XM_017020709.1 | XP_016876198.1 | 9 | 2877 | 136 | ||
| XM_017020710.1 | XP_016876199.1 | 10 | 2262 | 136 | ||
| XM_017020711.1 | XP_016876200.1 | 9 | 2163 | 136 | ||
| GenBank mRNA | AF318575.1 | - | 10 | 2058 | 136 | |
| AK001069.1 | - | 5 | 1442 | 136 | ||
| AK097027.1 | - | 6 | 2127 | 136 | ||
| BC008694.1 | - | 6 | 1578 | 136 | ||
| BC023569.1 | - | 9 | 2030 | 4127465 | 136 | |
| BQ008047.1 | - | 1 | 136 | |||
More Information
Set Membership: |
|

UPF3A
Genomic Map