Assay Details
| Gene Symbol: ARHGEF40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017021434.1 | XP_016876923.1 | 1 - 2 | 391 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017021434.1 transcripts.
Set Membership: |
|

ARHGEF40
Genomic Map