Assay Details
| Gene Symbol: HNRNPU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004501.3 | NP_004492.2 | 12 | 2421 | 88 | |
| NM_031844.2 | NP_114032.2 | 12 | 2478 | 88 | ||
| XM_017001115.1 | XP_016856604.1 | 12 | 2673 | 88 | ||
| XM_017001116.1 | XP_016856605.1 | 12 | 2478 | 88 | ||
| XM_017001117.1 | XP_016856606.1 | 12 | 2467 | 88 | ||
| GenBank mRNA | AF068846.1 | - | 12 | 2474 | 88 | |
| AF461014.1 | - | 9 | 1999 | 88 | ||
| AK126868.1 | - | 13 | 2219 | 88 | ||
| AK130669.1 | - | 11 | 1527 | 88 | ||
| AK297119.1 | - | 13 | 2312 | 88 | ||
| BC003367.1 | - | 12 | 2352 | 2900876 | 88 | |
| BC003621.2 | - | 12 | 2354 | 2966453 | 88 | |
| BC007950.2 | - | 12 | 1902 | 88 | ||
| BC015782.1 | - | 12 | 1955 | 4857112 | 88 | |
| BC024767.1 | - | 12 | 2354 | 4993703 | 88 | |
| BC034925.1 | - | 9 | 1345 | 88 | ||
| X65488.1 | - | 12 | 2244 | 88 | ||
More Information
Set Membership: |
|

HNRNPU
Genomic Map