Assay Details
| Gene Symbol: YY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003403.4 | NP_003394.1 | 1 | 142 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_003403.4 transcripts.
Set Membership: |
|

YY1
Genomic Map