Assay Details
| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011536443.1 | XP_011534745.1 | 2 - 3 | 813 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011536443.1 transcripts.
Set Membership: |
|

SLC25A29
Genomic Map