Assay Details
| Gene Symbol: LINC00221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027457.2 | 3 | 501 | 106 | ||
| GenBank mRNA | BX161407.1 | - | 2 | 304 | 106 | |
| BX248748.1 | - | 3 | 473 | 106 | ||
| BX356743.2 | - | 3 | 427 | 106 | ||
More Information
Set Membership: |
|

LINC00221
Genomic Map