Assay Details
| Gene Symbol: SCAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320778.1 | NP_001307707.1 | 10 | 1480 | 64 | |
| NM_005697.4 | NP_005688.2 | 9 | 1351 | 64 | ||
| XM_017021856.1 | XP_016877345.1 | 10 | 1405 | 64 | ||
| GenBank mRNA | AK096376.1 | - | 7 | 1497 | 64 | |
More Information
Set Membership: |
|

SCAMP2
Genomic Map