Assay Details
| Gene Symbol: SCAPER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017022282.1 | XP_016877771.1 | 21 | 3999 | 62 | |
| XM_017022283.1 | XP_016877772.1 | 21 | 3980 | 62 | ||
More Information
Set Membership: |
|

SCAPER
Genomic Map