Assay Details
| Gene Symbol: ALPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017022445.1 | XP_016877934.1 | 10 | 5367 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALPK3| Gene Symbol: ALPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017022445.1 | XP_016877934.1 | 10 | 5367 | 92 | |
Set Membership: |
|