Assay Details
| Gene Symbol: CHRFAM7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005254750.2 | XP_005254807.1 | 11 | 5615 | 146 | |
| XM_011522153.1 | XP_011520455.1 | 11 | 5140 | 146 | ||
| XM_011522155.2 | XP_011520457.1 | 9 | 5331 | 146 | ||
More Information
Set Membership: |
|

CHRFAM7A
Genomic Map