Assay Details
| Gene Symbol: ABHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007011.7 | NP_008942.3 | 15 | 8118 | 82 | |
| NM_152924.4 | NP_690888.1 | 11 | 7720 | 82 | ||
| XM_005254832.1 | XP_005254889.1 | 14 | 7756 | 82 | ||
| XM_005254833.1 | XP_005254890.1 | 11 | 7364 | 82 | ||
| XM_011521165.1 | XP_011519467.1 | 12 | 7569 | 82 | ||
| GenBank mRNA | AK095685.1 | - | 1 | 1483 | 82 | |
| CR749694.1 | - | 1 | 2966 | 82 | ||
More Information
Set Membership: |
|

ABHD2
Genomic Map