Assay Details
| Gene Symbol: SNX29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017023878.1 | XP_016879367.1 | 1 | 654 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017023878.1 transcripts.
Set Membership: |
|

SNX29
Genomic Map