Assay Details
| Gene Symbol: KAT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011545970.1 | XP_011544272.1 | 1 - 2 | 614 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011545970.1 transcripts.
Set Membership: |
|

KAT8
Genomic Map