Assay Details
| Gene Symbol: NRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005269444.4 | XP_005269501.1 | 10 | 5621 | 61 | |
| XM_011539172.2 | XP_011537474.1 | 10 | 5626 | 61 | ||
| XM_011539173.2 | XP_011537475.1 | 10 | 5623 | 61 | ||
| XM_011539175.2 | XP_011537477.1 | 9 | 5554 | 61 | ||
| XM_011539178.2 | XP_011537480.1 | 7 | 4812 | 61 | ||
| XM_017015573.1 | XP_016871062.1 | 11 | 5706 | 61 | ||
| XM_017015574.1 | XP_016871063.1 | 11 | 5701 | 61 | ||
| XM_017015575.1 | XP_016871064.1 | 11 | 5703 | 61 | ||
| XM_017015576.1 | XP_016871065.1 | 11 | 5698 | 61 | ||
| XM_017015577.1 | XP_016871066.1 | 10 | 5634 | 61 | ||
| XM_017015578.1 | XP_016871067.1 | 10 | 5629 | 61 | ||
| XM_017015579.1 | XP_016871068.1 | 10 | 5630 | 61 | ||
| XM_017015580.1 | XP_016871069.1 | 10 | 5626 | 61 | ||
| XM_017015581.1 | XP_016871070.1 | 9 | 5551 | 61 | ||
| XM_017015582.1 | XP_016871071.1 | 11 | 4976 | 61 | ||
| XM_017015583.1 | XP_016871072.1 | 10 | 4901 | 61 | ||
More Information
Set Membership: |
|

NRG3
Genomic Map