Assay Details
| Gene Symbol: CNTNAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017025238.1 | XP_016880727.1 | 1 - 2 | 442 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017025238.1 transcripts.
Set Membership: |
|

CNTNAP1
Genomic Map