Assay Details
| Gene Symbol: MED13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011525551.2 | XP_011523853.1 | 1 | 454 | 106 | |
| XM_011525552.2 | XP_011523854.1 | 1 | 454 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011525551.2, XM_011525552.2 transcripts.
Set Membership: |
|

MED13
Genomic Map